Anti-IgG F(c) Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
610-1122
ROCK610-1122EA 241 EUR
ROCK610-1122
Anti-IgG F(c) Goat Polyclonal Antibody
Antikörper
Anti-Mouse IgG F(c) antibody is suitable for ELISA, Western Blot and immunochemistry applications. Antibody should be optimized by end user for specific reactive conditions.

Recommended Dilutions: ELISA: 1:450000; Western Blot: 1:5000 - 1:50000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR