Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 649)

Lieferant: Rockland Immunochemicals
610-143-003
ROCK610-143-003EA 213 EUR
ROCK610-143-003
Anti-IgG F(c) Goat Polyclonal Antibody (DyLight® 649)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope: F(c)
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR