Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 800)

Lieferant: Rockland Immunochemicals
610-145-121
ROCK610-145-121EA 220 EUR
ROCK610-145-121
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 800)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR