Anti-IgG2b Goat Polyclonal Antibody (ATTO 532)

Lieferant: Rockland Immunochemicals
610-153-042
ROCK610-153-042EA 617 EUR
ROCK610-153-042
Anti-IgG2b Goat Polyclonal Antibody (ATTO 532)
Antikörper
Mouse IgG2b secondary antibody is suitable for ELISA.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: 1:4000 - 1:20000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: ATTO 532
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR