Anti-IgG (H+L) Goat Polyclonal Antibody (ATTO 532)

Lieferant: Rockland Immunochemicals
610-153-121S
ROCK610-153-121SEA 237 EUR
ROCK610-153-121S
Anti-IgG (H+L) Goat Polyclonal Antibody (ATTO 532)
Antikörper
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: ATTO 532
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR