Anti-IgG2a Goat Polyclonal Antibody (ATTO 594)
Lieferant: Rockland Immunochemicals
610-155-041
ROCK610-155-041EA 617 EUR
ROCK610-155-041
Anti-IgG2a Goat Polyclonal Antibody (ATTO 594)
Antikörper
Anti-Mouse IgG2a secondary antibody is suitable for ELISA.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: 1:4000 - 1:20000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: ATTO 594
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse