Anti-IgG2b Goat Polyclonal Antibody (ATTO 647N)
Lieferant: Rockland Immunochemicals
610-156-042
ROCK610-156-042EA 617 EUR
ROCK610-156-042
Anti-IgG2b Goat Polyclonal Antibody (ATTO 647N)
Antikörper
Anti-Mouse IgG2b secondary antibody is suitable for ELISA.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: 1:4000 - 1:20000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: ATTO 647N
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse