Anti-GDF15 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
610-401-B87S
ROCK610-401-B87SEA 226 EUR
ROCK610-401-B87S
Anti-GDF15 Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody is suitable for ELISA and western blotting of mouse and human NAG-1 Protein.
Recommended Dilutions: ELISA: 1:100000 - 1:120000; Western Blot: 1:3000 - 1:7000;
Type: Primary
Antigen: GDF15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Human