Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
610-401-C46
ROCK610-401-C46EA 341 EUR
ROCK610-401-C46
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody
Antikörper
Mouse IgG secondary antibody is available in a variety of formats.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Other Dilution: User Optimized
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse