Anti-IgG1/IgG2a/IgG2b/IgG3 gamma Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Lieferant: Rockland Immunochemicals
610-403-C46
ROCK610-403-C46EA 400 EUR
ROCK610-403-C46
Anti-IgG1/IgG2a/IgG2b/IgG3 gamma Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse