Anti-IgG3 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Rockland Immunochemicals
610-4043
ROCK610-4043EA 400 EUR
ROCK610-4043
Anti-IgG3 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Secondary Rabbit Anti-IgG3 Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG3 γ
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR