Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))

Lieferant: Rockland Immunochemicals
610-408-C46
ROCK610-408-C46EA 481 EUR
ROCK610-408-C46
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Antikörper
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR