Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
610-4111
ROCK610-4111EA 366 EUR
ROCK610-4111
Anti-lambda light chainlambda light chain Rabbit Polyclonal Antibody
Antikörper
Secondary Rabbit Anti-λ (lambda chain) Reacts with Mouse

Recommended Dilutions: ELISA: 1:40000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:3000;

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Light Chain
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR