Anti-IgG2b Rabbit Polyclonal Antibody (Peroxidase)
Lieferant: Rockland Immunochemicals
610-4342
ROCK610-4342EA 464 EUR
ROCK610-4342
Anti-IgG2b Rabbit Polyclonal Antibody (Peroxidase)
Antikörper
Anti-Mouse IgG2b peroxidase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:65000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG2b γ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse