Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
610-4507
ROCK610-4507EA 524 EUR
ROCK610-4507
Anti-IgM µ Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Anti-MOUSE IgM (alpha chain) (RABBIT) Antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000;
Do not freeze!

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: µ-chain
Host: Rabbit
Isotype: IgM
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR