Anti-IgG2a Rabbit Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
610-4641
ROCK610-4641EA 400 EUR
ROCK610-4641
Anti-IgG2a Rabbit Polyclonal Antibody (Biotin)
Antikörper
Secondary Rabbit Anti-IgG2a Reacts with Mouse
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG2a
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse