Anti-IgG3 Rabbit Polyclonal Antibody (Texas Red®)
Lieferant: Rockland Immunochemicals
610-4943
ROCK610-4943EA 400 EUR
ROCK610-4943
Anti-IgG3 Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Rabbit Anti-IgG3 Reacts with Mouse
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG3 γ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse