Anti-IgG (H+L) Rat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: Rockland Immunochemicals
611-500-122
ROCK611-500-122EA 421 EUR
ROCK611-500-122
Anti-IgG (H+L) Rat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit