Anti-IgG (H+L) Rat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
611-501-122
ROCK611-501-122EA 287 EUR
ROCK611-501-122
Anti-IgG (H+L) Rat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR