Anti-IgG (H+L) Rat Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
611-503-122
ROCK611-503-122EA 496 EUR
ROCK611-503-122
Anti-IgG (H+L) Rat Polyclonal Antibody (Peroxidase)
Antikörper
Suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunohistochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:10000-1:100000; Western Blot: 1:5000-1:40000; Immunochemistry: 1:500-1:5000; Other Dilution: User Optimized

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR