Anti-IgG (H+L) Rat Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
611-506-122
ROCK611-506-122EA 421 EUR
ROCK611-506-122
Anti-IgG (H+L) Rat Polyclonal Antibody (Biotin)
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Rat
Isotype:
Reactivity: Rabbit