Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 680)
Lieferant: Rockland Immunochemicals
611-744-127
ROCK611-744-127EA 220 EUR
ROCK611-744-127
Anti-IgG (H+L) Donkey Polyclonal Antibody (DyLight® 680)
Antikörper
The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000; Other Dilution: User Optimized
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Rabbit