Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
612-1303
ROCK612-1303EA 384 EUR
ROCK612-1303
Anti-IgG F(c) Goat Polyclonal Antibody (Peroxidase)
Antikörper
Secondary Goat Anti-IgG F(c) Reacts with Rat

Recommended Dilutions: ELISA: 1:130,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR