Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Lieferant: Rockland Immunochemicals
612-1325
ROCK612-1325EA 341 EUR
ROCK612-1325
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Antikörper
Secondary Goat Anti-IgG (H&L) Reacts with Rat
Recommended Dilutions: ELISA: 1:50000 - 1:350000; Western Blot: 1:5000 - 1:25000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rat