Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 649)
Lieferant: Rockland Immunochemicals
612-143-002
ROCK612-143-002EA 213 EUR
ROCK612-143-002
Anti-IgG (H+L) Goat Polyclonal Antibody (DyLight® 649)
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 100 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 649
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Rat