Anti-IgG F(c) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
612-1503
ROCK612-1503EA 421 EUR
ROCK612-1503
Anti-IgG F(c) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Secondary Goat Anti-IgG F(c) Reacts with Rat

Recommended Dilutions: ELISA: 1:2000 - 1:5000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR