Anti-IgG F(c) Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
612-1903
ROCK612-1903EA 384 EUR
ROCK612-1903
Anti-IgG F(c) Goat Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Goat Anti-IgG F(c) Reacts with Rat

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR