Anti-IgG (H+L) Donkey Polyclonal Antibody (Peroxidase)
Lieferant: Rockland Immunochemicals
613-703-168
ROCK613-703-168EA 464 EUR
ROCK613-703-168
Anti-IgG (H+L) Donkey Polyclonal Antibody (Peroxidase)
Antikörper
Anti-Sheep IgG (H&L) peroxidase conjugated antibody is suitable for immunoblotting (western or dot blot), ELISA, immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:65000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Sheep