Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
613-706-168
ROCK613-706-168EA 421 EUR
ROCK613-706-168
Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Antikörper
This product has been assayed against 1.0 µg of Sheep IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Sheep