Anti-IgM µ Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
618-101-007
ROCK618-101-007EA 308 EUR
ROCK618-101-007
Anti-IgM µ Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Ferret
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR