Anti-IgG Goat Polyclonal Antibody (Peroxidase)
Lieferant: Rockland Immunochemicals
618-103-012
ROCK618-103-012EA 468 EUR
ROCK618-103-012
Anti-IgG Goat Polyclonal Antibody (Peroxidase)
Antikörper
Secondary Goat Anti-IgG (gamma chain) Reacts with Ferret
Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Type: Secondary
Antigen: IgG γ
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: γ chain
Host: Goat
Isotype: IgG
Reactivity: Ferret