Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
618-106-130
ROCK618-106-130EA 341 EUR
ROCK618-106-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Biotin)
Antikörper
Secondary Goat Anti-IgG IgA IgM Reacts with Ferret

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Ferret
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR