Anti-FITC Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Lieferant: Rockland Immunochemicals
700-100-096
ROCK700-100-096EA 977 EUR
ROCK700-100-096
Anti-FITC Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: pan