Anti-Biotin Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
700-101-098
ROCK700-101-098EA 774 EUR
ROCK700-101-098
Anti-Biotin Goat Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Biotinylated BSA in a standard sandwich ELISA using Peroxidase conjugated Affinity Purified anti-Goat IgG (H&L) (Rabbit) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:1000-1:5000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:1000

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR