Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Lieferant: Rockland Immunochemicals
700-105-096
ROCK700-105-096EA 1030 EUR
ROCK700-105-096
Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.
Recommended Dilutions: ELISA: 1:38000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!
Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: pan