Anti-Biotin Goat Polyclonal Antibody (Cy2®)

Lieferant: Rockland Immunochemicals
700-111-098
ROCK700-111-098EA 1030 EUR
ROCK700-111-098
Anti-Biotin Goat Polyclonal Antibody (Cy2®)
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: Immunofluorescence: 1:1000-1:5000; Flow Cytometry: 1:500-1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: Cy2®
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR