Anti-IgG (H+L) Rabbit Polyclonal Antibody (Cy3®)

Lieferant: Rockland Immunochemicals
708-404-002
ROCK708-404-002EA 504 EUR
ROCK708-404-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Cy3®)
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 250 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Horse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR