Anti-lambda light chain Goat Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
709-1311
ROCK709-1311EA 526 EUR
ROCK709-1311
Anti-lambda light chain Goat Polyclonal Antibody (Peroxidase)
Antikörper
Secondary Goat Anti-λ lambda chain Reacts with Human

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR