Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
710-109-019
ROCK710-109-019EA 562 EUR
ROCK710-109-019
Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)
Antikörper
Recommended Dilutions:
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR