Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
712-1622
ROCK712-1622EA 400 EUR
ROCK712-1622
Anti-IgG F(c) Goat Polyclonal Antibody (Biotin)
Antikörper
This product has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using Peroxidase Conjugated Streptavidin and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR