Anti-IgM µ Goat Polyclonal Antibody (PE (Phycoerythrin))
Lieferant: Rockland Immunochemicals
712-1807
ROCK712-1807EA 650 EUR
ROCK712-1807
Anti-IgM µ Goat Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.
Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!
Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rat