Anti-IgG (H+L) Donkey Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: Rockland Immunochemicals
712-708-149
ROCK712-708-149EA 650 EUR
ROCK712-708-149
Anti-IgG (H+L) Donkey Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR