Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)
Lieferant: Rockland Immunochemicals
713-709-168
ROCK713-709-168EA 617 EUR
ROCK713-709-168
Anti-IgG (H+L) Donkey Polyclonal Antibody (Texas Red®)
Antikörper
Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Donkey
Isotype:
Reactivity: Sheep