Anti-Cyclin Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
K911
ROCKK911EA 1850 EUR
ROCKK911
Anti-Cyclin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Cyclin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR