Immunoblotting kits, MaxTag™
Lieferant: Rockland Immunochemicals
These accurate and sensitive kits are ideal for Western blotting or dot blotting methods. They offer flexibility with two unique substrates: Bright Blue TMB substrate and Stable Brown DAB substrate. Components are colour-coded for error-free results. With MaxTag™ you can run up to 40 (10×10cm) immunoblots.
- Rapid
- Accurate
- Flexible with two unique substrates
Lieferumfang: Complete kit includes ultra pure Tween® 20, IgG Bovine Serum Albumin, biotinylated anti-IgG antibody, enhanced Steptavadin-Peroxidase conjugate, TMB substrate and precipitating agent, DAB substrate tablets, hydrogen peroxide (H₂O₂) solution, reusable dropper bottles, control IgG, and instruction manual.