Western Blotting-Blockierungsreagenz, Amersham ECL™
Lieferant: Cytiva
RPN2125
RPN2125EA 89.2 EUR
RPN2125
Western Blotting-Blockierungsreagenz, Amersham ECL™
Elektrophorese-Reagenzien Western Blotting Reagents
Amersham ECL Blocking Agent is for blocking nitrocellulose and PVDF membranes to be used with Amersham ECL, Amersham ECL Prime and Amersham ECL Select Western Blotting Detection Reagents
Can be used with the Amersham ECL Western blotting detection family. This blocking agent is intended for blocking nitrocellulose and PVDF membranes used with ECL and ECL Plus Western Blotting Detection Reagents.
Lieferumfang: Jede Packung enthält 40 g des Membran-Blockierungsreagenz, ausreichend für mindestens 20 Miniblots.