Precast horizontal gel kits, SERVALYT™ PRECOTES™ Tf Kit
Lieferant: Serva
42803.01
SERA42803.01EA 0 EUR
SERA42803.01
Precast horizontal gel kits, SERVALYT™ PRECOTES™ Tf Kit
Elektrophorese-Gele Fertiggele
For transferrin (Tf) analysis by isoelectric focusing.
SERVALYT™ PRECOTES™ Tf Kit contains:
5× SERVALYT™ PRECOTES™ Tf gels, 245×125 mm, 300 µm
Anode and cathode buffer solutions
Applicator strips and electrode wicks