Dialysierschläuche
Lieferant: Serva
44139.01 44139.02 44144.01 44144.02 44145.01 44145.04 44146.01 44146.04 44148.01 44148.02
SERA44139.01EA 82.6 EUR
SERA44139.01 SERA44139.02 SERA44144.01 SERA44144.02 SERA44145.01 SERA44145.04 SERA44146.01 SERA44146.04 SERA44148.01 SERA44148.02
Dialysierschläuche
Schläuche Dialyseschläuche
Dialysis tubing is made of cellulose membrane. It contains low amounts of impurities such as sulphides and heavy metals.
  • High chemical and physical resistance
  • pH range: 2 to 12
  • Temperature range: 4 to 60 °C