PEEK tubing
Lieferant: Trajan Scientific and Medical
1302015008-10F 1301005001-5F 1301007002-5F 1302020004-10F 1302030005-10F 1302030001-10F 1301010003-5F 1301015008-5F 1301020004-5F 1302004006-5F 1302005001-5F 1302007002-5F 1302010003-5F 1302013004-5F 1302015008-5F 1302020004-5F 1302030005-5F 1302030001-5F 1302040000-5F 1303062000-5F 1303080000-5F 1304002000-5F 1304003006-5F 1304004001-5F 1304005005-5F 1304006002-5F 1301005001-10F 1301007002-10F 1301010003-10F 1301015008-10F 1301020004-10F 1302004006-10F 1302005001-10F 1302007002-10F 1302010003-10F 1302013004-10F 1302040000-10F 1303062000-10F 1303080000-10F 1304002000-10F 1304003006-10F 1304004001-10F 1304005005-10F 1304006002-10F 1301005001-50F 1301007002-50F 1301010003-50F 1301015008-50F 1301020004-50F 1302004006-50F 1302005001-50F 1302007002-50F 1302010003-50F 1302013004-50F 1302015008-50F 1302020004-50F 1302030005-50F 1302030001-50F 1302040000-50F 1303062000-50F 1303080000-50F 1304002000-50F 1304003006-50F 1304004001-50F 1304005005-50F 1304006002-50F 1301005001100F 1301007002100F 1301015008100F 1301020004100F 1302004006100F 1302005001100F 1302007002100F 1302010003100F 1302013004100F 1302015008100F 1302020004100F 1302030005100F 1302030001100F 1302040000100F 1303062000100F 1303080000100F 1304002000100F 1304003006100F 1304004001100F 1304005005100F 1304006002100F
SGEA1302015008-10FEA 40.9 EUR
SGEA1302015008-10F SGEA1301005001-5F SGEA1301007002-5F SGEA1302020004-10F SGEA1302030005-10F SGEA1302030001-10F SGEA1301010003-5F SGEA1301015008-5F SGEA1301020004-5F SGEA1302004006-5F SGEA1302005001-5F SGEA1302007002-5F SGEA1302010003-5F SGEA1302013004-5F SGEA1302015008-5F SGEA1302020004-5F SGEA1302030005-5F SGEA1302030001-5F SGEA1302040000-5F SGEA1303062000-5F SGEA1303080000-5F SGEA1304002000-5F SGEA1304003006-5F SGEA1304004001-5F SGEA1304005005-5F SGEA1304006002-5F SGEA1301005001-10F SGEA1301007002-10F SGEA1301010003-10F SGEA1301015008-10F SGEA1301020004-10F SGEA1302004006-10F SGEA1302005001-10F SGEA1302007002-10F SGEA1302010003-10F SGEA1302013004-10F SGEA1302040000-10F SGEA1303062000-10F SGEA1303080000-10F SGEA1304002000-10F SGEA1304003006-10F SGEA1304004001-10F SGEA1304005005-10F SGEA1304006002-10F SGEA1301005001-50F SGEA1301007002-50F SGEA1301010003-50F SGEA1301015008-50F SGEA1301020004-50F SGEA1302004006-50F SGEA1302005001-50F SGEA1302007002-50F SGEA1302010003-50F SGEA1302013004-50F SGEA1302015008-50F SGEA1302020004-50F SGEA1302030005-50F SGEA1302030001-50F SGEA1302040000-50F SGEA1303062000-50F SGEA1303080000-50F SGEA1304002000-50F SGEA1304003006-50F SGEA1304004001-50F SGEA1304005005-50F SGEA1304006002-50F SGEA1301005001100F SGEA1301007002100F SGEA1301015008100F SGEA1301020004100F SGEA1302004006100F SGEA1302005001100F SGEA1302007002100F SGEA1302010003100F SGEA1302013004100F SGEA1302015008100F SGEA1302020004100F SGEA1302030005100F SGEA1302030001100F SGEA1302040000100F SGEA1303062000100F SGEA1303080000100F SGEA1304002000100F SGEA1304003006100F SGEA1304004001100F SGEA1304005005100F SGEA1304006002100F
PEEK tubing
Schläuche Chromatographie-Schläuche
Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
  • Chemical resistant and inert to most commonly used solvents
  • Bio-inert and bio-compatible
  • Colour coded to industry standards for easy identification of inner diameter
  • Leach-free colorants
  • Smooth inner walls and precise IDs
  • Works well with polymer and stainless steel fittings
  • Maximum operating temperature of 100 °C
PEEK tubing is a flexible alternative to stainless steel tubing in high pressure applications. Also ensures reproducibility in chromatographic flow rates through smooth internal surface along with consistent and tight toleranced IDs.
Bestellinformation: Accessories should be ordered separately.