Anti-Her2 / ERBB2 Rabbit Polyclonal Antibody

Lieferant: Sino Biological
50714-T48-200
SINO50714-T48-200EA 0 EUR
SINO50714-T48-200
Anti-Her2 / ERBB2 Rabbit Polyclonal Antibody
Antikörper
Anti-Her2 / ERBB2 antibody

Type: Primary
Antigen: Her2/ERBB2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR